missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse CCL12 / MCP-5 Protein
A DNA sequence encoding the Mus musculus (Mouse) CCL12 / MCP-5, was expressed in the hosts and tags indicated.
245.00€ - 3405.00€
Specifications
Accession Number | NP_035461.2 |
---|---|
Gene ID (Entrez) | 20293 |
Name | CCL12 / MCP-5 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16035751
|
enQuireBio™
QP8648-EC-100UG |
100 μg |
770.00€
100µg |
Estimated Shipment: 19-06-2024
Log in to see stock. |
|||||
16055751
|
enQuireBio™
QP8648-EC-1MG |
1 mg |
3405.00€
1mg |
Estimated Shipment: 19-06-2024
Log in to see stock. |
|||||
16045751
|
enQuireBio™
QP8648-EC-10UG |
10 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16065751
|
enQuireBio™
QP8648-EC-200UG |
200 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16075751
|
enQuireBio™
QP8648-EC-500UG |
500 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16085751
|
enQuireBio™
QP8648-EC-50UG |
50 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
Specifications
NP_035461.2 | |
CCL12 / MCP-5 Protein | |
Not determined. | |
Full Length | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |
20293 | |
Research Use Only | |
Ccl12 | |
Recombinant Protein | |
E. coli | |
GPDAVSTPVT CCYNVVKQKI HVRKLKSYRR ITSSQCPREA VIFRTILDKE ICADPKEKWV KNSINHLDKT SQTFILEPSC LG | |
Greater than 90% as determined by SDS-PAGE. |