missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Angiopoietin-like protein 1
A DNA sequence encoding the Mus musculus (Mouse) Angiopoietin-like protein 8, was expressed in the hosts and tags indicated.
245.00€ - 2645.00€
Specifications
Accession Number | NP_001074409.1 |
---|---|
Gene ID (Entrez) | 624219 |
Name | Angiopoietin-like protein 1 |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16091611
|
enQuireBio™
QP7923-YE-100UG |
100 μg |
770.00€
100µg |
Estimated Shipment: 26-06-2024
Log in to see stock. |
|||||
16031621
|
enQuireBio™
QP7923-YE-500UG |
500 μg |
2195.00€
500µg |
Estimated Shipment: 26-06-2024
Log in to see stock. |
|||||
16001621
|
enQuireBio™
QP7923-YE-10UG |
10 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16021621
|
enQuireBio™
QP7923-YE-200UG |
200 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16041621
|
enQuireBio™
QP7923-YE-50UG |
50 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16011621
|
enQuireBio™
QP7923-YE-1MG |
1 mg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
Specifications
NP_001074409.1 | |
Angiopoietin-like protein 1 | |
Not determined. | |
Full Length | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |
624219 | |
Research Use Only | |
Gm6484 | |
Recombinant Protein | |
Yeast | |
VRPAPVAPLG GPEPAQYEEL TLLFHGALQL GQALNGVYRA TEARLTEAGH SLGLYDRALE FLGTEVRQGQ DATQELRTSL SEIQVEEDAL HLRAEATARS LGEVARAQQA LRDTVRRLQV QLRGAWLGQA HQEFETLKAR ADKQSHLLWA LTGHVQRQQR EMAEQQQWLR QIQQRLHTAA LPA | |
Greater than 90% as determined by SDS-PAGE. |