missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Mouse ADAM10 Protein
Click to view available options
Quantity:
20 μg
Unit Size:
20µg
Description
The Recombinant Mouse ADAM10 Protein is derived from Sf 21 (baculovirus). The Recombinant Mouse ADAM10 Protein has been validated for the following applications: Enzyme Activity.
Specifications
Specifications
| Formulation | Lyophilized from a 0.2 μm filtered solution in MES, NaCl, CaCl 2 ZnCl 2 and Brij-35. |
| Gene ID (Entrez) | 11487 |
| Molecular Weight (g/mol) | 52 kDa |
| Name | ADAM10 |
| Quantity | 20 μg |
| Source | Spodoptera frugiperda,Sf 21 (baculovirus)-derived mouse ADAM10 protein Thr215-Glu673,with a C-terminal 10-His tag |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20 to -70° C as supplied. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
| Structural Form | Recombinant Mouse ADAM10 is prone to proteolytic cleavage at C-terminus. The predominant form of the purified protein lacks the His tag. |
| Conjugate | Unconjugated |
| Recombinant | Recombinant |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction