missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Mouse Activin B Protein
Click to view available options
Formulation:
Lyophilized from a 0.2μm filtered solution in HCl with BSA as a carrier protein.
Lyophilized from a 0.2μm filtered solution in HCl.
Unit Size:
10µg
Description
The Recombinant Mouse Activin B Protein is derived from HEK293. The Recombinant Mouse Activin B Protein has been validated for the following applications: Binding Activity.
Order Info
The product is shipped at ambient temperature. Upon receipt, store it immediately at the temperature recommended below.
Specifications
Specifications
| Accession Number | Q04999 |
| Concentration | Reconstitute at 100μg/mL in sterile 4mM HCl. |
| Formulation | Lyophilized from a 0.2μm filtered solution in HCl. |
| Molecular Weight (g/mol) | 13 kDa (monomer) |
| Name | Activin B, Carrier Free |
| Quantity | 10 μg |
| Source | Human embryonic kidney cell,HEK293-derived mouse Activin B protein Gly297-Ala411 |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
| Gene Alias | activin AB beta polypeptide, Activin beta-B chain, INHBB, inhibin beta B chain, inhibin, beta B, inhibin, beta B (activin AB beta polypeptide), Inhibin, beta-2 |
| Conjugate | Unconjugated |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction