missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human X-ray repair cross-complementing protein 1

A DNA sequence encoding the Homo sapiens (Human) X-ray repair cross-complementing protein 5, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6894-ec-10ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16026610

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance


Description

Description

Specifications

Specifications

NP_066964.1
X-ray repair cross-complementing protein 1
Research Use Only
XRCC5
Recombinant Protein
E. coli
LTIGSNLSIR IAAYKSILQE RVKKTWTVVD AKTLKKEDIQ KETVYCLNDD DETEVLKEDI IQGFRYGSDI VPFSKVDEEQ MKYKSEGKCF SVLGFCKSSQ VQRRFFMGNQ VLKVFAARDD EAAAVALSSL IHALDDLDMV AIVRYAYDKR ANPQVGVAFP HIKHNYECLV YVQLPFMEDL RQYMFSSLKN SKKYAPTEAQ LNAVD
Greater than 90% as determined by SDS-PAGE.
7520
10 μg
Not determined.
Partial (see sequence information for more details).
Human
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers