missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human X-ray repair cross-complementing protein 1
A DNA sequence encoding the Homo sapiens (Human) X-ray repair cross-complementing protein 5, was expressed in the hosts and tags indicated.
160.00€ - 2385.00€
Specifications
Accession Number | NP_066964.1 |
---|---|
Gene ID (Entrez) | 7520 |
Name | X-ray repair cross-complementing protein 1 |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16016610
|
enQuireBio™
QP6894-EC-100UG |
100 μg |
530.00€
100µg |
Estimated Shipment: 21-06-2024
Log in to see stock. |
|||||
16036610
|
enQuireBio™
QP6894-EC-1MG |
1 mg |
2385.00€
1mg |
Estimated Shipment: 21-06-2024
Log in to see stock. |
|||||
16026610
|
enQuireBio™
QP6894-EC-10UG |
10 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16046610
|
enQuireBio™
QP6894-EC-200UG |
200 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16056610
|
enQuireBio™
QP6894-EC-500UG |
500 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16066610
|
enQuireBio™
QP6894-EC-50UG |
50 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
Specifications
NP_066964.1 | |
X-ray repair cross-complementing protein 1 | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
7520 | |
Research Use Only | |
XRCC5 | |
Recombinant Protein | |
E. coli | |
LTIGSNLSIR IAAYKSILQE RVKKTWTVVD AKTLKKEDIQ KETVYCLNDD DETEVLKEDI IQGFRYGSDI VPFSKVDEEQ MKYKSEGKCF SVLGFCKSSQ VQRRFFMGNQ VLKVFAARDD EAAAVALSSL IHALDDLDMV AIVRYAYDKR ANPQVGVAFP HIKHNYECLV YVQLPFMEDL RQYMFSSLKN SKKYAPTEAQ LNAVD | |
Greater than 90% as determined by SDS-PAGE. |