missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human VTI1B Protein
A DNA sequence encoding the Homo sapiens (Human) VTI1B, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8206-ec-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_006361.1 | |
VTI1B Protein | |
Research Use Only | |
VTI1B | |
Recombinant Protein | |
E. coli | |
SSEHFEKLHE IFRGLHEDLQ GVPERLLGTA GTEEKKKLIR DFDEKQQEAN ETLAEMEEEL RYAPLSFRNP MMSKLRNYRK DLAKLHREVR STPLTATPGG RGDMKYGIYA VENEHMNRLQ SQRAMLLQGT ESLNRATQSI ERSHRIATET DQIGSEIIEE LGEQRDQLER TKSRLVNTSE NLSKSRKILR SMSRKVTTNK LL | |
Greater than 90% as determined by SDS-PAGE. |
10490 | |
500 μg | |
Not determined. | |
Cytoplasmic Domain | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |