missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human VTI1B Protein
A DNA sequence encoding the Homo sapiens (Human) VTI1B, was expressed in the hosts and tags indicated.
160.00€ - 2385.00€
Specifications
Accession Number | NP_006361.1 |
---|---|
Gene ID (Entrez) | 10490 |
Name | VTI1B Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16023271
|
enQuireBio™
QP8206-EC-100UG |
100 μg |
530.00€
100µg |
Estimated Shipment: 17-06-2024
Log in to see stock. |
|||||
16043271
|
enQuireBio™
QP8206-EC-1MG |
1 mg |
2385.00€
1mg |
Estimated Shipment: 17-06-2024
Log in to see stock. |
|||||
16033271
|
enQuireBio™
QP8206-EC-10UG |
10 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16053271
|
enQuireBio™
QP8206-EC-200UG |
200 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16063271
|
enQuireBio™
QP8206-EC-500UG |
500 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16073271
|
enQuireBio™
QP8206-EC-50UG |
50 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
Specifications
NP_006361.1 | |
VTI1B Protein | |
Not determined. | |
Cytoplasmic Domain | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
10490 | |
Research Use Only | |
VTI1B | |
Recombinant Protein | |
E. coli | |
SSEHFEKLHE IFRGLHEDLQ GVPERLLGTA GTEEKKKLIR DFDEKQQEAN ETLAEMEEEL RYAPLSFRNP MMSKLRNYRK DLAKLHREVR STPLTATPGG RGDMKYGIYA VENEHMNRLQ SQRAMLLQGT ESLNRATQSI ERSHRIATET DQIGSEIIEE LGEQRDQLER TKSRLVNTSE NLSKSRKILR SMSRKVTTNK LL | |
Greater than 90% as determined by SDS-PAGE. |