Learn More
R&D Systems™ Recombinant Human VEGF 111 Protein
Product Code. 18376377
Click to view available options
Formulation:
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein.
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA.
Unit Size:
10µg
Product Code. 18376377
Brand: R&D Systems™ 5336VE010
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.
The Recombinant Human VEGF 111 Protein is derived from E. coli. The Recombinant Human VEGF 111 Protein has been validated for the following applications: Bioactivity.
TRUSTED_SUSTAINABILITY
Specifications
Formulation Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein.
Gene ID (Entrez) 7422
Molecular Weight (g/mol) 13 kDa
Name VEGF 111
Quantity 10 μg
Source E. coli-derived human VEGF protein Ala27-Arg137,with an N-terminal Met & Pro28-Arg137
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Structural Form Disulfide-linked homodimer
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.