missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Upstream stimulatory factor 1 Protein
A DNA sequence encoding the Homo sapiens (Human) Upstream stimulatory factor 1, was expressed in the hosts and tags indicated.
160.00€ - 2385.00€
Specifications
Accession Number | NP_001263302.1 |
---|---|
Gene ID (Entrez) | 7391 |
Name | Upstream stimulatory factor 1 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16096440
|
enQuireBio™
QP6872-EC-100UG |
100 μg |
530.00€
100µg |
Estimated Shipment: 21-06-2024
Log in to see stock. |
|||||
16016450
|
enQuireBio™
QP6872-EC-1MG |
1 mg |
2385.00€
1mg |
Estimated Shipment: 21-06-2024
Log in to see stock. |
|||||
16006450
|
enQuireBio™
QP6872-EC-10UG |
10 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16026450
|
enQuireBio™
QP6872-EC-200UG |
200 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16036450
|
enQuireBio™
QP6872-EC-500UG |
500 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16046450
|
enQuireBio™
QP6872-EC-50UG |
50 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
Specifications
NP_001263302.1 | |
Upstream stimulatory factor 1 Protein | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
7391 | |
Research Use Only | |
USF1 | |
Recombinant Protein | |
E. coli | |
GQQKTAETEE GTVQIQEGAV ATGEDPTSVA IASIQSAATF PDPNVKYVFR TENGGQVMYR VIQVSEGQLD GQTEGTGAIS GYPATQSMTQ AVIQGAFTSD DAVDTEGTAA ETHYTYFPST AVGDGAGGTT SGSTAAVVTT QGSEALLGQA TPPGTGQFFV MMSPQEVLQG GSQRSIAPRT HPYSPKSEAP RTTRDEKRRA QHNEVERRRR DKINNWIVQL SKIIPDCSME STKSGQSKGG ILSKACDYIQ ELRQSNHRLS EELQGLDQLQ LDNDVLRQQV EDLKNKNLLL RAQLRHHGLE VVIKNDSN | |
Greater than 90% as determined by SDS-PAGE. |