missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Upstream stimulatory factor 1 Protein

A DNA sequence encoding the Homo sapiens (Human) Upstream stimulatory factor 1, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6872-ec-500ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16036450

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance


Description

Description

Specifications

Specifications

NP_001263302.1
Upstream stimulatory factor 1 Protein
Research Use Only
USF1
Recombinant Protein
E. coli
GQQKTAETEE GTVQIQEGAV ATGEDPTSVA IASIQSAATF PDPNVKYVFR TENGGQVMYR VIQVSEGQLD GQTEGTGAIS GYPATQSMTQ AVIQGAFTSD DAVDTEGTAA ETHYTYFPST AVGDGAGGTT SGSTAAVVTT QGSEALLGQA TPPGTGQFFV MMSPQEVLQG GSQRSIAPRT HPYSPKSEAP RTTRDEKRRA QHNEVERRRR DKINNWIVQL SKIIPDCSME STKSGQSKGG ILSKACDYIQ ELRQSNHRLS EELQGLDQLQ LDNDVLRQQV EDLKNKNLLL RAQLRHHGLE VVIKNDSN
Greater than 90% as determined by SDS-PAGE.
7391
500 μg
Not determined.
Partial (see sequence information for more details).
Human
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers