Learn More
enQuireBio™ Recombinant Human Upstream stimulatory factor 1 Protein
A DNA sequence encoding the Homo sapiens (Human) Upstream stimulatory factor 1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6872-ec-10ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001263302.1 | |
Upstream stimulatory factor 1 Protein | |
Research Use Only | |
USF1 | |
Recombinant Protein | |
E. coli | |
GQQKTAETEE GTVQIQEGAV ATGEDPTSVA IASIQSAATF PDPNVKYVFR TENGGQVMYR VIQVSEGQLD GQTEGTGAIS GYPATQSMTQ AVIQGAFTSD DAVDTEGTAA ETHYTYFPST AVGDGAGGTT SGSTAAVVTT QGSEALLGQA TPPGTGQFFV MMSPQEVLQG GSQRSIAPRT HPYSPKSEAP RTTRDEKRRA QHNEVERRRR DKINNWIVQL SKIIPDCSME STKSGQSKGG ILSKACDYIQ ELRQSNHRLS EELQGLDQLQ LDNDVLRQQV EDLKNKNLLL RAQLRHHGLE VVIKNDSN | |
Greater than 90% as determined by SDS-PAGE. |
7391 | |
10 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |