missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human UDP-glucose 6-dehydrogenase Protein
A DNA sequence encoding the Homo sapiens (Human) UDP-glucose 6-dehydrogenase, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6869-ec-10ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001171629.1 | |
UDP-glucose 6-dehydrogenase Protein | |
Research Use Only | |
UGDH | |
Recombinant Protein | |
E. coli | |
MFEIKKICCI GAGYVGGPTC SVIAHMCPEI RVTVVDVNES RINAWNSPTL PIYEPGLKEV VESCRGKNLF FSTNIDDAIK EADLVFISVN TPTKTYGMGK GRAADLKYIE ACARRIVQNS NGYKIVTEKS TVPVRAAESI RRIFDANTKP NLNLQVLSNP EFLAEGTAIK DLKNPDRVLI GGDETPEGQR AVQALCAVYE HWVPREKILT TNTWSSELSK LAANAFLAQR ISSINSISAL CEATGADVEE VATAIGMDQR IGNKFLKASV GFGGSCFQKD VLNLVYLCEA LNLPEVARYW QQVIDMNDYQ RRRFASRIID SLFNTVTDKK IAILGFAFKK DTGDTRESSS IYISKYLMDE GAHLHIYDPK VPREQIVVDL SHPGVSEDDQ VSRLVTISKD PYEACDGAHA VVICTEWDMF KELDYERIHK KMLKPAFIFD GRRVLDGLHN ELQTIGFQIE TIGKKVSSKR IPYAPSGEIP KFSLQDPPNK KPKV | |
Greater than 90% as determined by SDS-PAGE. |
7358 | |
10 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |