missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human UDP-glucose 6-dehydrogenase Protein
A DNA sequence encoding the Homo sapiens (Human) UDP-glucose 6-dehydrogenase, was expressed in the hosts and tags indicated.
160.00€ - 2385.00€
Specifications
Accession Number | NP_001171629.1 |
---|---|
Gene ID (Entrez) | 7358 |
Name | UDP-glucose 6-dehydrogenase Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16016430
|
enQuireBio™
QP6869-EC-100UG |
100 μg |
530.00€
100µg |
Estimated Shipment: 21-06-2024
Log in to see stock. |
|||||
16036430
|
enQuireBio™
QP6869-EC-1MG |
1 mg |
2385.00€
1mg |
Estimated Shipment: 21-06-2024
Log in to see stock. |
|||||
16026430
|
enQuireBio™
QP6869-EC-10UG |
10 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16046430
|
enQuireBio™
QP6869-EC-200UG |
200 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16056430
|
enQuireBio™
QP6869-EC-500UG |
500 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16066430
|
enQuireBio™
QP6869-EC-50UG |
50 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
Specifications
NP_001171629.1 | |
UDP-glucose 6-dehydrogenase Protein | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
7358 | |
Research Use Only | |
UGDH | |
Recombinant Protein | |
E. coli | |
MFEIKKICCI GAGYVGGPTC SVIAHMCPEI RVTVVDVNES RINAWNSPTL PIYEPGLKEV VESCRGKNLF FSTNIDDAIK EADLVFISVN TPTKTYGMGK GRAADLKYIE ACARRIVQNS NGYKIVTEKS TVPVRAAESI RRIFDANTKP NLNLQVLSNP EFLAEGTAIK DLKNPDRVLI GGDETPEGQR AVQALCAVYE HWVPREKILT TNTWSSELSK LAANAFLAQR ISSINSISAL CEATGADVEE VATAIGMDQR IGNKFLKASV GFGGSCFQKD VLNLVYLCEA LNLPEVARYW QQVIDMNDYQ RRRFASRIID SLFNTVTDKK IAILGFAFKK DTGDTRESSS IYISKYLMDE GAHLHIYDPK VPREQIVVDL SHPGVSEDDQ VSRLVTISKD PYEACDGAHA VVICTEWDMF KELDYERIHK KMLKPAFIFD GRRVLDGLHN ELQTIGFQIE TIGKKVSSKR IPYAPSGEIP KFSLQDPPNK KPKV | |
Greater than 90% as determined by SDS-PAGE. |