missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Tyrosine-protein kinase JAK1 Protein

A DNA sequence encoding the Homo sapiens (Human) Tyrosine-protein kinase JAK1, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6244-ec-10ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16021880

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance


Description

Description

Specifications

Specifications

NP_001307852.1
Tyrosine-protein kinase JAK1 Protein
Research Use Only
JAK1
Recombinant Protein
E. coli
EQNPDIVSEK KPATEVDPTH FEKRFLKRIR DLGEGHFGKV ELCRYDPEGD NTGEQVAVKS LKPESGGNHI ADLKKEIEIL RNLYHENIVK YKGICTEDGG NGIKLIMEFL PSGSLKEYLP KNKNKINLKQ QLKYAVQICK GMDYLGSRQY VHRDLAARNV LVESEHQVKI GDFGLTKAIE TDKEYYTVKD DRDSPVFWYA PECLMQSKFY IASDVWSFGV TLHELLTYCD SDSSPMALFL KMIGPTHGQM TVTRLVNTLK EGKRLPCPPN CPDEVYQLMR KCWEFQPSNR TSFQNLIEGF EALLK
Greater than 90% as determined by SDS-PAGE.
3716
10 μg
Not determined.
Partial (see sequence information for more details).
Human
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers