missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Tyrosine-protein kinase JAK1 Protein
A DNA sequence encoding the Homo sapiens (Human) Tyrosine-protein kinase JAK1, was expressed in the hosts and tags indicated.
245.00€ - 3405.00€
Specifications
Accession Number | NP_001307852.1 |
---|---|
Gene ID (Entrez) | 3716 |
Name | Tyrosine-protein kinase JAK1 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16011880
|
enQuireBio™
QP6244-EC-100UG |
100 μg |
770.00€
100µg |
Estimated Shipment: 14-06-2024
Log in to see stock. |
|||||
16031880
|
enQuireBio™
QP6244-EC-1MG |
1 mg |
3405.00€
1mg |
Estimated Shipment: 14-06-2024
Log in to see stock. |
|||||
16021880
|
enQuireBio™
QP6244-EC-10UG |
10 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16041880
|
enQuireBio™
QP6244-EC-200UG |
200 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16051880
|
enQuireBio™
QP6244-EC-500UG |
500 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16061880
|
enQuireBio™
QP6244-EC-50UG |
50 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
Specifications
NP_001307852.1 | |
Tyrosine-protein kinase JAK1 Protein | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
3716 | |
Research Use Only | |
JAK1 | |
Recombinant Protein | |
E. coli | |
EQNPDIVSEK KPATEVDPTH FEKRFLKRIR DLGEGHFGKV ELCRYDPEGD NTGEQVAVKS LKPESGGNHI ADLKKEIEIL RNLYHENIVK YKGICTEDGG NGIKLIMEFL PSGSLKEYLP KNKNKINLKQ QLKYAVQICK GMDYLGSRQY VHRDLAARNV LVESEHQVKI GDFGLTKAIE TDKEYYTVKD DRDSPVFWYA PECLMQSKFY IASDVWSFGV TLHELLTYCD SDSSPMALFL KMIGPTHGQM TVTRLVNTLK EGKRLPCPPN CPDEVYQLMR KCWEFQPSNR TSFQNLIEGF EALLK | |
Greater than 90% as determined by SDS-PAGE. |