missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Tyrosine-protein kinase ABL1 Protein
A DNA sequence encoding the Homo sapiens (Human) Tyrosine-protein kinase ABL1, was expressed in the hosts and tags indicated.
160.00€ - 2385.00€
Specifications
Accession Number | NP_005148.2 |
---|---|
Gene ID (Entrez) | 25 |
Name | Tyrosine-protein kinase ABL1 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16004821
|
enQuireBio™
QP8475-EC-100UG |
100 μg |
530.00€
100µg |
Estimated Shipment: 25-06-2024
Log in to see stock. |
|||||
16024821
|
enQuireBio™
QP8475-EC-1MG |
1 mg |
2385.00€
1mg |
Estimated Shipment: 25-06-2024
Log in to see stock. |
|||||
16014821
|
enQuireBio™
QP8475-EC-10UG |
10 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16034821
|
enQuireBio™
QP8475-EC-200UG |
200 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16044821
|
enQuireBio™
QP8475-EC-500UG |
500 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16054821
|
enQuireBio™
QP8475-EC-50UG |
50 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
Specifications
NP_005148.2 | |
Tyrosine-protein kinase ABL1 Protein | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |
25 | |
Research Use Only | |
ABL | |
Recombinant Protein | |
E. coli | |
ICLKLVGCKS KKGLSSSSSC YLEEALQRPV ASDFEPQGLS EAARWNSKEN LLAGPSENDP NLFVALYDFV ASGDNTLSIT KGEKLRVLGY NHNGEWCEAQ TKNGQGWVPS NYITPVNSLE KHSWYHGPVS RNAAEYLLSS GINGSFLVRE SESSPGQRSI SLRYEGRVYH YRINTASDGK LYVSSESRFN T | |
Greater than 90% as determined by SDS-PAGE. |