missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Tyrosine-protein kinase ABL1 Protein
A DNA sequence encoding the Homo sapiens (Human) Tyrosine-protein kinase ABL1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8475-ec-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_005148.2 | |
Tyrosine-protein kinase ABL1 Protein | |
Research Use Only | |
ABL | |
Recombinant Protein | |
E. coli | |
ICLKLVGCKS KKGLSSSSSC YLEEALQRPV ASDFEPQGLS EAARWNSKEN LLAGPSENDP NLFVALYDFV ASGDNTLSIT KGEKLRVLGY NHNGEWCEAQ TKNGQGWVPS NYITPVNSLE KHSWYHGPVS RNAAEYLLSS GINGSFLVRE SESSPGQRSI SLRYEGRVYH YRINTASDGK LYVSSESRFN T | |
Greater than 90% as determined by SDS-PAGE. |
25 | |
500 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |