missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Recombinant Human Tyrosine Hydroxylase His Protein
Shop All Bio Techne Products
Click to view available options
:
0.01mg; Unlabeled
Description
Recombinant Tyrosine Hydroxylase protein with an N-terminal His tag and corresponding to amino acids 1 - 497. Source: E. coli (Accession #: P07101-2) The Recombinant Human Tyrosine Hydroxylase His Protein is derived from E. coli. The Recombinant Human Tyrosine Hydroxylase His Protein has been validated for the following applications: Western Blot, Immunohistochemistry-Paraffin, SDS-Page.
Specifications
Specifications
| Accession Number | P07101-2 |
| For Use With (Application) | Immunohistochemistry-Paraffin, SDS-PAGE, Western Blot |
| Formulation | 300 mM NaCl, 2.7 mM KCl, 4.3 mM Na2HPO4, 1.47 mM KH2PO4, 5% glycerol, 1 mM DTT, pH 7.4 |
| Gene ID (Entrez) | 7054 |
| Molecular Weight (g/mol) | M.W. theroretical: 60 kDa |
| Name | Tyrosine Hydroxylase Protein |
| Storage Requirements | −80°C. Avoid freeze-thaw cycles. |
| Cross Reactivity | Human |
| Research Category | Cancer, Lipid and Metabolism, Neurodegeneration, Neuronal Cell Markers, Neuroscience, Neurotransmission, Phospho Specific, Prostate Cancer |
| Purity or Quality Grade | >95% |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction