missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Trimethylguanosine synthase Protein
A DNA sequence encoding the Homo sapiens (Human) Trimethylguanosine synthase, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7949-ec-10ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001304831.1 | |
Trimethylguanosine synthase Protein | |
Research Use Only | |
TGS1 | |
Recombinant Protein | |
E. coli | |
MRVIAIDIDP VKIALARNNA EVYGIADKIE FICGDFLLLA SFLKADVVFL SPPWGGPDYA TAETFDIRTM MSPDGFEIFR LSKKITNNIV YFLPRNADID QVASLAGPGG QVEIEQNFLN NKLKTITAYF GDLIRRPASE T | |
Greater than 90% as determined by SDS-PAGE. |
96764 | |
10 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |