missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Trimethylguanosine synthase Protein
A DNA sequence encoding the Homo sapiens (Human) Trimethylguanosine synthase, was expressed in the hosts and tags indicated.
160.00€ - 2385.00€
Specifications
Accession Number | NP_001304831.1 |
---|---|
Gene ID (Entrez) | 96764 |
Name | Trimethylguanosine synthase Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16031791
|
enQuireBio™
QP7949-EC-100UG |
100 μg |
530.00€
100µg |
Estimated Shipment: 17-06-2024
Log in to see stock. |
|||||
16051791
|
enQuireBio™
QP7949-EC-1MG |
1 mg |
2385.00€
1mg |
Estimated Shipment: 17-06-2024
Log in to see stock. |
|||||
16041791
|
enQuireBio™
QP7949-EC-10UG |
10 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16061791
|
enQuireBio™
QP7949-EC-200UG |
200 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16071791
|
enQuireBio™
QP7949-EC-500UG |
500 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16081791
|
enQuireBio™
QP7949-EC-50UG |
50 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
Specifications
NP_001304831.1 | |
Trimethylguanosine synthase Protein | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
96764 | |
Research Use Only | |
TGS1 | |
Recombinant Protein | |
E. coli | |
MRVIAIDIDP VKIALARNNA EVYGIADKIE FICGDFLLLA SFLKADVVFL SPPWGGPDYA TAETFDIRTM MSPDGFEIFR LSKKITNNIV YFLPRNADID QVASLAGPGG QVEIEQNFLN NKLKTITAYF GDLIRRPASE T | |
Greater than 90% as determined by SDS-PAGE. |