missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human TOP1 Protein

Product Code. 15994689
Click to view available options
Quantity:
1 mg
20 μg
5 μg
Unit Size:
1mg
20µg
5µg
This item is not returnable. View return policy

Product Code. 15994689

Brand: enQuireBio™ QP137835ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A cDNA sequence encoding the TOP1 was constructed and used to recombinantly synthesize the protein.

Specifications

Gene ID (Entrez) 7150
Name TOP1 Protein
Quantity 5 μg
Regulatory Status Research Use Only
Endotoxin Concentration Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Gene Symbol TOP1
Product Type Recombinant Protein
Cross Reactivity Human
Species Baculovirus
Protein Tag His
Sequence MSGDHLHNDS QIEADFRLND SHKHKDKHKD REHRHKEHKK EKDREKSKHS NSEHKDSEKK HKEKEKTKHK DGSSEKHKDK HKDRDKEKRK EEKVRASGDA KIKKEKENGF SSPPQIKDEP EDDGYFVPPK EDIKPLKRPR DEDDADYKPK KIKTEDTKKE KKRKLEEEED GKLKKPKNKD KDKKVPEPDN KKKKPKKEEE QKWKWWEEER YPEGIKWKFL EHKGPVFAPP YEPLPENVKF YYDGKVMKLS PKAEEVATFF AKMLDHEYTT KEIFRKNFFK DWRKEMTNEE KNIITNLSKC DFTQMSQYFK AQTEARKQMS KEEKLKIKEE NEKLLKEYGF CIMDNHKERI ANFKIEPPGL FRGRGNHPKM GMLKRRIMPE DIIINCSKDA KVPSPPPGHK WKEVRHDNKV TWLVSWTENI QGSIKYIMLN PSSRIKGEKD WQKYETARRL KKCVDKIRNQ YREDWKSKEM KVRQRAVALY FIDKLALRAG NEKEEGETAD TVGCCSLRVE HINLHPELDG QEYVVEFDFL GKDSIRYYNK VPVEKRVFKN LQLFMENKQP EDDLFDRLNT GILNKHLQDL MEGLTAKVFR TYNASITLQQ QLKELTAPDE NIPAKILSYN RANRAVAILC NHQRAPPKTF EKSMMNLQTK IDAKKEQLAD ARRDLKSAKA DAKVMKDAKT KKVVESKKKA VQRLEEQLMK LEVQATDREE NKQIALGTSK LNYLDPRITV AWCKKWGVPI EKIYNKTQRE KFAWAIDMAD EDYEF
Buffer TOP1 is supplied in 16mM HEPES buffer pH 7.5, 400mM sodium chloride, and 20% glycerol.
Purity or Quality Grade Greater than 95% as determined by SDS-PAGE.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.