missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human TIMP-1 Western Blot Standard Protein
Click to view available options
Quantity:
1 Vial
Unit Size:
100µL
Description
The Recombinant Human TIMP-1 Western Blot Standard Protein has been validated for the following applications: Western Blot.
Specifications
Specifications
| Gene ID (Entrez) | 7076 |
| Name | TIMP-1 Western Blot Standard |
| Quantity | 1 Vial |
| Storage Requirements | Store the unopened product at -20 to -70° C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Conjugate | Unconjugated |
| Recombinant | Recombinant |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction