missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human TIMM21 Protein

A DNA sequence encoding the Homo sapiens (Human) TIMM21, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8108-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16042701

  • 2385.00€ / 1mg
Estimated Shipment: 25-06-2024
to see stock.
Add to basket

Description

Description

Specifications

Specifications

NP_054896.2
TIMM21 Protein
Research Use Only
TIMM21
Recombinant Protein
E. coli
MICTFLRAVQ YTEKLHRSSA KRLLLPYIVL NKACLKTEPS LRCGLQYQKK TLRPRCILGV TQKTIWTQGP SPRKAKEDGS KQVSVHRSQR GGTAVPTSQK VKEAGRDFTY LIVVLFGISI TGGLFYTIFK ELFSSSSPSK IYGRALEKCR SHPEVIGVFG ESVKGYGEVT RRGRRQHVRF TEYVKDGLKH TCVKFYIEGS EPGKQGTVYA QVKENPGSGE YDFRYIFVEI ESYPRRTIII EDNRSQDD
Greater than 80% as determined by SDS-PAGE.
29090
1 mg
Not determined.
Full Length
Human
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers