missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human TIMM21 Protein
A DNA sequence encoding the Homo sapiens (Human) TIMM21, was expressed in the hosts and tags indicated.
160.00€ - 2385.00€
Specifications
Accession Number | NP_054896.2 |
---|---|
Gene ID (Entrez) | 29090 |
Name | TIMM21 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16022701
|
enQuireBio™
QP8108-EC-100UG |
100 μg |
530.00€
100µg |
Estimated Shipment: 28-05-2024
Log in to see stock. |
|||||
16042701
|
enQuireBio™
QP8108-EC-1MG |
1 mg |
2385.00€
1mg |
Estimated Shipment: 28-05-2024
Log in to see stock. |
|||||
16032701
|
enQuireBio™
QP8108-EC-10UG |
10 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16052701
|
enQuireBio™
QP8108-EC-200UG |
200 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16062701
|
enQuireBio™
QP8108-EC-500UG |
500 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16072701
|
enQuireBio™
QP8108-EC-50UG |
50 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
Specifications
NP_054896.2 | |
TIMM21 Protein | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |
29090 | |
Research Use Only | |
TIMM21 | |
Recombinant Protein | |
E. coli | |
MICTFLRAVQ YTEKLHRSSA KRLLLPYIVL NKACLKTEPS LRCGLQYQKK TLRPRCILGV TQKTIWTQGP SPRKAKEDGS KQVSVHRSQR GGTAVPTSQK VKEAGRDFTY LIVVLFGISI TGGLFYTIFK ELFSSSSPSK IYGRALEKCR SHPEVIGVFG ESVKGYGEVT RRGRRQHVRF TEYVKDGLKH TCVKFYIEGS EPGKQGTVYA QVKENPGSGE YDFRYIFVEI ESYPRRTIII EDNRSQDD | |
Greater than 80% as determined by SDS-PAGE. |