missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Thrombospondin-1 Protein
A DNA sequence encoding the Homo sapiens (Human) Thrombospondin-1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8570-ec-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_003237.2 | |
Thrombospondin-1 Protein | |
Research Use Only | |
THBS1 | |
Recombinant Protein | |
E. coli | |
NRIPESGGDN SVFDIFELTG AARKGSGRRL VKGPDPSSPA FRIEDANLIP PVPDDKFQDL VDAVRAEKGF LLLASLRQMK KTRGTLLALE RKDHSGQVFS VVSNGKAGTL DLSLTVQGKQ HVVSVEEALL ATGQWKSITL FVQEDRAQLY IDCEKMENAE LDVPIQSVFT RDLASIARLR IAKGGVNDNF QGVLQNVRFV FGTTPEDILR NKGCSSSTSV LLTLDNNVVN GSSPAIRTNY IGHKTKDLQA ICGISCDELS SMVLELRGLR TIVTTLQDSI RKVTEENKEL ANELRRPPLC YHNGVQYRNN EEWTVDSCTE CHCQNSVTIC KK | |
Greater than 90% as determined by SDS-PAGE. |
7057 | |
500 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |