missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Thrombospondin-1 Protein
A DNA sequence encoding the Homo sapiens (Human) Thrombospondin-1, was expressed in the hosts and tags indicated.
160.00€ - 2385.00€
Specifications
Accession Number | NP_003237.2 |
---|---|
Gene ID (Entrez) | 7057 |
Name | Thrombospondin-1 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16025341
|
enQuireBio™
QP8570-EC-100UG |
100 μg |
530.00€
100µg |
Estimated Shipment: 21-06-2024
Log in to see stock. |
|||||
16045341
|
enQuireBio™
QP8570-EC-1MG |
1 mg |
2385.00€
1mg |
Estimated Shipment: 21-06-2024
Log in to see stock. |
|||||
16035341
|
enQuireBio™
QP8570-EC-10UG |
10 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16055341
|
enQuireBio™
QP8570-EC-200UG |
200 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16065341
|
enQuireBio™
QP8570-EC-500UG |
500 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16075341
|
enQuireBio™
QP8570-EC-50UG |
50 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
Specifications
NP_003237.2 | |
Thrombospondin-1 Protein | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
7057 | |
Research Use Only | |
THBS1 | |
Recombinant Protein | |
E. coli | |
NRIPESGGDN SVFDIFELTG AARKGSGRRL VKGPDPSSPA FRIEDANLIP PVPDDKFQDL VDAVRAEKGF LLLASLRQMK KTRGTLLALE RKDHSGQVFS VVSNGKAGTL DLSLTVQGKQ HVVSVEEALL ATGQWKSITL FVQEDRAQLY IDCEKMENAE LDVPIQSVFT RDLASIARLR IAKGGVNDNF QGVLQNVRFV FGTTPEDILR NKGCSSSTSV LLTLDNNVVN GSSPAIRTNY IGHKTKDLQA ICGISCDELS SMVLELRGLR TIVTTLQDSI RKVTEENKEL ANELRRPPLC YHNGVQYRNN EEWTVDSCTE CHCQNSVTIC KK | |
Greater than 90% as determined by SDS-PAGE. |