missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Threonine--tRNA ligase, mitochondrial Protein
A DNA sequence encoding the Homo sapiens (Human) Threonine--tRNA ligase, mitochondrial, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6762-ec-10ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001258825.1 | |
Threonine--tRNA ligase, mitochondrial Protein | |
Research Use Only | |
TARS2 | |
Recombinant Protein | |
E. coli | |
EHYQEDMFAV QPPGSDRPPS SQSDDSTRHI TDTLALKPMN CPAHCLMFAH RPRSWRELPL RLADFGALHR AEASGGLGGL TRLRCFQQDD AHIFCTTDQL EAEIQSCLDF LRSVYAVLGF SFRLALSTRP SGFLGDPCLW DQAEQVLKQA LKEFGEPWDL NSGDGAFYGP KIDVHLHDAL GRPHQCGTIQ LDFQLPLRFD LQYKGQAGAL ERPVLIHRAV LGSVERLLGV LAESCGGKWP LWLSPFQVVV IPVGSEQEEY AKEAQQSLRA AGLVSDLDAD SGLTLSRRIR RAQLAHYNFQ FVVGQKEQSK RTVNIRTRDN RRLGEWDLPE AVQRLVELQN TRVPNAEEIF | |
Greater than 90% as determined by SDS-PAGE. |
80222 | |
10 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |