missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Threonine--tRNA ligase, mitochondrial Protein
A DNA sequence encoding the Homo sapiens (Human) Threonine--tRNA ligase, mitochondrial, was expressed in the hosts and tags indicated.
160.00€ - 2385.00€
Specifications
Accession Number | NP_001258825.1 |
---|---|
Gene ID (Entrez) | 80222 |
Name | Threonine--tRNA ligase, mitochondrial Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16015650
|
enQuireBio™
QP6762-EC-100UG |
100 μg |
530.00€
100µg |
Estimated Shipment: 28-05-2024
Log in to see stock. |
|||||
16035650
|
enQuireBio™
QP6762-EC-1MG |
1 mg |
2385.00€
1mg |
Estimated Shipment: 28-05-2024
Log in to see stock. |
|||||
16025650
|
enQuireBio™
QP6762-EC-10UG |
10 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16045650
|
enQuireBio™
QP6762-EC-200UG |
200 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16055650
|
enQuireBio™
QP6762-EC-500UG |
500 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16065650
|
enQuireBio™
QP6762-EC-50UG |
50 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
Specifications
NP_001258825.1 | |
Threonine--tRNA ligase, mitochondrial Protein | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
80222 | |
Research Use Only | |
TARS2 | |
Recombinant Protein | |
E. coli | |
EHYQEDMFAV QPPGSDRPPS SQSDDSTRHI TDTLALKPMN CPAHCLMFAH RPRSWRELPL RLADFGALHR AEASGGLGGL TRLRCFQQDD AHIFCTTDQL EAEIQSCLDF LRSVYAVLGF SFRLALSTRP SGFLGDPCLW DQAEQVLKQA LKEFGEPWDL NSGDGAFYGP KIDVHLHDAL GRPHQCGTIQ LDFQLPLRFD LQYKGQAGAL ERPVLIHRAV LGSVERLLGV LAESCGGKWP LWLSPFQVVV IPVGSEQEEY AKEAQQSLRA AGLVSDLDAD SGLTLSRRIR RAQLAHYNFQ FVVGQKEQSK RTVNIRTRDN RRLGEWDLPE AVQRLVELQN TRVPNAEEIF | |
Greater than 90% as determined by SDS-PAGE. |