Learn More
R&D Systems™ Recombinant Human TGF-beta 3 (CHO-expressed) Protein
Product Code. 18767566
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
Product Code. 18767566
Brand: R&D Systems™ 8420B3100/CF
Measured by its ability to inhibit the IL-4-dependent proliferation of HT-2 mouse T cells. The ED50 for this effect is 0.01 to 0.04 ng/mL.
TRUSTED_SUSTAINABILITY
Specifications
Accession Number P10600
For Use With (Application) Bioactivity
Gene ID (Entrez) 7043
Molecular Weight (g/mol) 13 kDa
Name ARVD, ARVD1, FLJ16571, LDS5, RNHF, TGFB3, TGFbeta 3, TGF-beta3, TGF-beta-3, transforming growth factor beta-3, transforming growth factor, beta 3
Quantity 100 μg
Source Chinese Hamster Ovary cell line, CHO-derived human TGF-beta 3 protein Ala301-Ser412
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 3 months, -20 to -70 °C under sterile conditions after reconstitution.
Gene Symbol TGFB3
Conjugate Unconjugated
Species Human
Reconstitution Reconstitute at 50 μg/mL in 4 mM HCl.
Buffer Lyophilized from a 0.2 μm filtered solution in Acetonitrile and TFA.
Endotoxin Level <0.10 EU per 1 μg of the protein by the LAL method.
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.