missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human TGF-beta 1 (CHO-Expressed) Protein, CF

Product Code. 18765965
Change view
Click to view available options
Quantity:
1 mg
10 μg
100 μg
50 μg
500 μg
Unit Size:
100µg
10µg
1mg
500µg
50µg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18765965 10 μg 10µg
18765785 500 μg 500µg
18746255 100 μg 100µg
18755735 1 mg 1mg
18705935 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 18765965 Supplier R&D Systems™ Supplier No. 11409BH010

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Measured by its ability to inhibit the IL-4-dependent proliferation of HT-2 mouse T cells. Tsang, M. et al. (1995) Cytokine 7:389. The ED50 for this effect is 0.025-0.200 ng/mL.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P01137.2
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in Acetonitrile and TFA with Trehalose.
Gene ID (Entrez) 7040
Molecular Weight (g/mol) M.W. (Observed): 12 kDa, under reducing conditions & 24 kDa under non-reducing conditions., M.W. (Theroretical): 13 kDa
Quantity 10 μg
Source Chinese Hamster Ovary cell line, CHO-derived human TGF-beta 1 protein Ala279-Ser390
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU per 1 μg of the protein by the LAL method.
Gene Alias CEDLAP, DPD1, latency-associated peptide, TGF beta, TGFB, TGFbeta, TGF-beta 1 protein, TGF-beta-1, transforming growth factor beta-1, transforming growth factor, beta 1
Conjugate Unconjugated
Reconstitution Reconstitute the 10 μg size at 100 μg/mL in 4 mM HCl. Reconstitute all other sizes at 250 μg/mL in 4 mM HCl.
Purity or Quality Grade >97%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein TGF-beta 1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.