missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Recombinant Human TFB2M His Protein

Product Code. 18157961 Shop All Bio Techne Products
Click to view available options
Quantity:
0.05 mg
250 μg
Unit Size:
0.05mg
250µg
This item is not returnable. View return policy

Product Code. 18157961

Brand: Novus Biologicals™ NBP222878

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Highly purified. Generating reliable and reproducible results.

A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 20-396 of Human TFB2M The Recombinant Human TFB2M Protein is derived from E. coli. The Recombinant Human TFB2M Protein has been validated for the following applications: SDS-Page.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number NP_071761
For Use With (Application) SDS-PAGE
Formulation 20mM Tris-HCl buffer (pH 8.0), 0.15 M NaCl, 10% glycerol, 1mM DTT
Gene ID (Entrez) 64216
Molecular Weight (g/mol) 45.8kDa
Name TFB2M Protein
Purification Method SDS-PAGE
Quantity 0.05 mg
Source E. Coli
Immunogen TFB2M, 20-396aa, Human, His tag, E.coli MGSSHHHHHH SSGLVPRGSH MGSMAGRFCI LGSEAATRKH LPARNHCGLS DSSPQLWPEP DFRNPPRKAS KASLDFKRYV TDRRLAETLA QIYLGKPSRP PHLLLECNPG PGILTQALLE AGAKVVALES DKTFIPHLES LGKNLDGKLR VIHCDFFKLD PRSGGVIKPP AMSSRGLFKN LGIEAVPWTA DIPLKVVGMF PSRGEKRALW KLAYDLYSCT SIYKFGRIEV NMFIGEKEFQ KLMADPGNPD LYHVLSVIWQ LACEIKVLHM EPWSSFDIYT RKGPLENPKR RELLDQLQQK LYLIQMIPRQ NLFTKNLTPM NYNIFFHLLK HCFGRRSATV IDHLRSLTPL DARDILMQIG KQEDEKVVNM HPQDFKTLFE TIERSKDCAY KWLYDETLED R
Storage Requirements −20°C or −80°C. Avoid freeze-thaw cycles.
Regulatory Status RUO
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Research Category metabolism
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.