missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human T-cell receptor alpha chain C region Protein

A DNA sequence encoding the Homo sapiens (Human) T-cell receptor alpha chain C region, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6836-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16096170

  • 476.00€ / 100µg

Estimated Shipment: 04-08-2023
to see stock.
Add to basket

Description

Description

Specifications

Specifications

T-cell receptor alpha chain C region Protein
100 μg
Not determined.
Full Length
Human
His
Tris-based buffer, 50% glycerol
Greater than 90% as determined by SDS-PAGE.
Research Use Only
TRAC
Recombinant Protein
E. coli
PNIQNPDPAV YQLRDSKSSD KSVCLFTDFD SQTNVSQSKD SDVYITDKTV LDMRSMDFKS NSAVAWSNKS DFACANAFNN SIIPEDTFFP SPESSCDVKL VEKSFETDTN LNFQNLSVIG FRILLLKVAG FNLLMTLRLW SS
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers