missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Synaptosomal-associated protein 29 Protein
A DNA sequence encoding the Homo sapiens (Human) Synaptosomal-associated protein 29, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6703-ec-200ug
Additional Details : Weight : 0.01000kg
Specifications
NP_004773.1 | |
Synaptosomal-associated protein 29 Protein | |
Research Use Only | |
SNAP29 | |
Recombinant Protein | |
E. coli | |
MSAYPKSYNP FDDDGEDEGA RPAPWRDARD LPDGPDAPAD RQQYLRQEVL RRAEATAAST SRSLALMYES EKVGVASSEE LARQRGVLER TEKMVDKMDQ DLKISQKHIN SIKSVFGGLV NYFKSKPVET PPEQNGTLTS QPNNRLKEAI STSKEQEAKY QASHPNLRKL DDTDPVPRGA GSAMSTDAYP KNPHLRAYHQ KIDSNLDELS MGLGRLKDIA LGMQTEIEEQ DDILDRLTTK VDKLDVNIKS TERKVRQL | |
Greater than 80% as determined by SDS-PAGE. |
9342 | |
200 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |