missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Synaptosomal-associated protein 29 Protein
A DNA sequence encoding the Homo sapiens (Human) Synaptosomal-associated protein 29, was expressed in the hosts and tags indicated.
160.00€ - 2385.00€
Specifications
Accession Number | NP_004773.1 |
---|---|
Gene ID (Entrez) | 9342 |
Name | Synaptosomal-associated protein 29 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16035240
|
enQuireBio™
QP6703-EC-100UG |
100 μg |
530.00€
100µg |
Estimated Shipment: 28-05-2024
Log in to see stock. |
|||||
16055240
|
enQuireBio™
QP6703-EC-1MG |
1 mg |
2385.00€
1mg |
Estimated Shipment: 28-05-2024
Log in to see stock. |
|||||
16045240
|
enQuireBio™
QP6703-EC-10UG |
10 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16065240
|
enQuireBio™
QP6703-EC-200UG |
200 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16075240
|
enQuireBio™
QP6703-EC-500UG |
500 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16085240
|
enQuireBio™
QP6703-EC-50UG |
50 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
Specifications
NP_004773.1 | |
Synaptosomal-associated protein 29 Protein | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |
9342 | |
Research Use Only | |
SNAP29 | |
Recombinant Protein | |
E. coli | |
MSAYPKSYNP FDDDGEDEGA RPAPWRDARD LPDGPDAPAD RQQYLRQEVL RRAEATAAST SRSLALMYES EKVGVASSEE LARQRGVLER TEKMVDKMDQ DLKISQKHIN SIKSVFGGLV NYFKSKPVET PPEQNGTLTS QPNNRLKEAI STSKEQEAKY QASHPNLRKL DDTDPVPRGA GSAMSTDAYP KNPHLRAYHQ KIDSNLDELS MGLGRLKDIA LGMQTEIEEQ DDILDRLTTK VDKLDVNIKS TERKVRQL | |
Greater than 80% as determined by SDS-PAGE. |