missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human STOM / EPB72 Protein

A DNA sequence encoding the Homo sapiens (Human) STOM / EPB72, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6740-ec-200ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16045500

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance


Description

Description

Specifications

Specifications

NP_004090.4
STOM / EPB72 Protein
Research Use Only
STOM
Recombinant Protein
E. coli
MAEKRHTRDS EAQRLPDSFK DSPSKGLGPC GWILVAFSFL FTVITFPISI WMCIKIIKEY ERAIIFRLGR ILQGGAKGPG LFFILPCTDS FIKVDMRTIS FDIPPQEILT KDSVTISVDG VVYYRVQNAT LAVANITNAD SATRLLAQTT LRNVLGTKNL SQILSDREEI AHNMQSTLDD ATDAWGIKVE RVEIKDVKLP VQLQRAMAAE AEASREARAK VIAAEGEMNA SRALKEASMV ITESPAALQL RYLQTLTTIA AEKNSTIVFP LPIDMLQGII GAKHSHLG
Greater than 90% as determined by SDS-PAGE.
2040
200 μg
Not determined.
Full Length
Human
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers