missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human STOM / EPB72 Protein
A DNA sequence encoding the Homo sapiens (Human) STOM / EPB72, was expressed in the hosts and tags indicated.
245.00€ - 3405.00€
Specifications
Accession Number | NP_004090.4 |
---|---|
Gene ID (Entrez) | 2040 |
Name | STOM / EPB72 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16015500
|
enQuireBio™
QP6740-EC-100UG |
100 μg |
770.00€
100µg |
Estimated Shipment: 21-06-2024
Log in to see stock. |
|||||
16035500
|
enQuireBio™
QP6740-EC-1MG |
1 mg |
3405.00€
1mg |
Estimated Shipment: 21-06-2024
Log in to see stock. |
|||||
16025500
|
enQuireBio™
QP6740-EC-10UG |
10 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16045500
|
enQuireBio™
QP6740-EC-200UG |
200 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16055500
|
enQuireBio™
QP6740-EC-500UG |
500 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16065500
|
enQuireBio™
QP6740-EC-50UG |
50 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
Specifications
NP_004090.4 | |
STOM / EPB72 Protein | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
2040 | |
Research Use Only | |
STOM | |
Recombinant Protein | |
E. coli | |
MAEKRHTRDS EAQRLPDSFK DSPSKGLGPC GWILVAFSFL FTVITFPISI WMCIKIIKEY ERAIIFRLGR ILQGGAKGPG LFFILPCTDS FIKVDMRTIS FDIPPQEILT KDSVTISVDG VVYYRVQNAT LAVANITNAD SATRLLAQTT LRNVLGTKNL SQILSDREEI AHNMQSTLDD ATDAWGIKVE RVEIKDVKLP VQLQRAMAAE AEASREARAK VIAAEGEMNA SRALKEASMV ITESPAALQL RYLQTLTTIA AEKNSTIVFP LPIDMLQGII GAKHSHLG | |
Greater than 90% as determined by SDS-PAGE. |