Learn More
R&D Systems™ Recombinant Human SorLA (aa 82-753) Protein
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
Description
The Recombinant Human SorLA (aa 82-753) Protein is derived from HEK293. The Recombinant Human SorLA (aa 82-753) Protein has been validated for the following applications: Bioactivity.
Specifications
Specifications
| Accession Number | Q92673 |
| For Use With (Application) | Bioactivity |
| Formulation | Lyophilized from a 0.2 μm filtered solution in PBS. |
| Gene ID (Entrez) | 6653 |
| Molecular Weight (g/mol) | 76 kDa |
| Quantity | 50 μg |
| Source | Human embryonic kidney cell, HEK293-derived human SorLA protein Ser82-Pro753, with a C-terminal 6-His tag |
| Storage Requirements | 12 months from date of receipt, ≤ -20°C as supplied; 1 month, 2 to 8°C under sterile conditions after reconstitution; 3 months, ≤ -20°C under sterile conditions after reconstitution. |
| Regulatory Status | RUO |
| Endotoxin Concentration | <0.10 EU per 1 μg of the protein by the LAL method. |
| Show More |