missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human Sonic Hedgehog/Shh (C24II) N-Terminus

Product Code. 18717726
Change view
Click to view available options
Quantity:
100 μg
25 μg
500 μg
Formulation:
Lyophilized from a 0.2 μm filtered solution in PBS and NaCl with BSA as a carrier protein.
Lyophilized from a 0.2μm filtered solution in PBS and NaCl with BSA as a carrier protein.
Lyophilized from a 0.2μm filtered solution in PBS and NaCl.
Unit Size:
100µg
25µg
500µg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Formulation Unit Size
18717726 500 μg Lyophilized from a 0.2 μm filtered solution in PBS and NaCl with BSA as a carrier protein. 500µg
18166580 25 μg Lyophilized from a 0.2μm filtered solution in PBS and NaCl with BSA as a carrier protein. 25µg
18301718 100 μg Lyophilized from a 0.2μm filtered solution in PBS and NaCl with BSA as a carrier protein. 100µg
18198060 25 μg Lyophilized from a 0.2μm filtered solution in PBS and NaCl. 25µg
18106680 100 μg Lyophilized from a 0.2μm filtered solution in PBS and NaCl. 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 18717726 Supplier R&D Systems™ Supplier No. 1845SH500
  • ONLINE EXCLUSIVE PRICE
    2590.00€
    2445.45€ / 500µg
    Save 144.55€
    5% Off
 Restricted Item
Estimated Shipment: 28-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

The Recombinant Human Sonic Hedgehog/Shh (C24II) N-Terminus is derived from E. coli. The Recombinant Human Sonic Hedgehog/Shh (C24II) N-Terminus has been validated for the following applications: Bioactivity.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number NP_000184.1
Formulation Lyophilized from a 0.2 μm filtered solution in PBS and NaCl with BSA as a carrier protein.
Gene ID (Entrez) 6469
Molecular Weight (g/mol) 20 kDa
Name Sonic Hedgehog/Shh (C24II), N-Terminus
Quantity 500 μg
Source E. coli-derived human Sonic Hedgehog/Shh protein Cys24-Gly197 (Cys24Ile-Ile),with an N-terminal Met
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Gene Alias HHG1, HHG-1, HLP3, HPE3, MCOPCB5, MCOPCB5sonic hedgehog (Drosophila) homolog, Shh, ShhNC, SMMCI, SMMCIsonic hedgehog homolog (Drosophila), sonic hedgehog, sonic hedgehog homolog, sonic hedgehog protein, TPT, TPTPS
Conjugate Unconjugated
Recombinant Recombinant
Reconstitution Reconstitute at 100 to 200 μg/mL in sterile PBS containing at least 0.1% human or bovine serum albumin, and allow up to 24 hours at 2°C to 8°C for complete reconstitution.
Product Line R&D Systems
Purity or Quality Grade 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.