missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human SNRNP25 Protein
Specifications
Specifications
| Name | SNRNP25 Protein |
| Quantity | 1 mg |
| Regulatory Status | Research Use Only |
| Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
| Product Type | Recombinant Protein |
| Cross Reactivity | Human |
| Species | E. coli |
| Protein Tag | His |
| Sequence | MGSSHHHHHH SSGLVPRGSH MDVFQEGLAM VVQDPLLCDL PIQVTLEEVN SQIALEYGQA MTVRVCKMDG EVMPVVVVQS ATVLDLKKAI QRYVQLKQER EGGIQHISWS YVWRTYHLTS AGEKLTEDRK KLRDYGIRNR DEVSFIKKLR QK |
| Buffer | SNRNP25 protein solution (0.5 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 20% glycerol, 2mM DTT and 0.2M NaCl. |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?