missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human SMARCB1 Protein
A DNA sequence encoding the Homo sapiens (Human) SMARCB1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7605-ec-100ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001007469.1 | |
SMARCB1 Protein | |
100 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
6598 | |
Greater than 90% as determined by SDS-PAGE. | |
Research Use Only | |
SMARCB1 | |
Recombinant Protein | |
E. coli | |
MMMALSKTFG QKPVKFQLED DGEFYMIGSE VGNYLRMFRG SLYKRYPSLW RRLATVEERK KIVASSHGKK TKPNTKDHGY TTLATSVTLL KASEVEEILD GNDEKYKAVS ISTEPPTYLR EQKAKRNSQW VPTLPNSSHH LDAVPCSTTI NRNRMGRDKK RTFPLCFDDH DPAVIHENAS QPEVLVPIRL DMEIDGQKLR DAFTWNMNEK LMTPEMFSEI LCDDLDLNPL TFVPAIASAI RQQIESYPTD SILEDQSDQR VIIKLNIHVG NISLVDQFEW DMSEKENSPE KFALKLCSEL GLGGEFVTTI AYSIRGQLSW HQKTYAFSEN PLPTVEIAIR NTGDADQWCP LLETLTDAEM EKKIRDQDRN TRRMR |