missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human SMARCB1 Protein

Product Code. p-7154276
Click to view available options
Quantity:
1 mg
10 μg
100 μg
200 μg
50 μg
500 μg
Unit Size:
100µg
10µg
1mg
200µg
500µg
50µg
This item is not returnable. View return policy

Product Code. 16009710

Brand: enQuireBio™ QP7605ec10ug

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products

This item is not returnable. View return policy

A DNA sequence encoding the Homo sapiens (Human) SMARCB1, was expressed in the hosts and tags indicated.

Specifications

Accession Number NP_001007469.1
Gene ID (Entrez) 6598
Name SMARCB1 Protein
Quantity 10 μg
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
Gene Symbol SMARCB1
Protein Length Partial (see sequence information for more details).
Product Type Recombinant Protein
Cross Reactivity Human
Species E. coli
Protein Tag His-SUMO
Sequence MMMALSKTFG QKPVKFQLED DGEFYMIGSE VGNYLRMFRG SLYKRYPSLW RRLATVEERK KIVASSHGKK TKPNTKDHGY TTLATSVTLL KASEVEEILD GNDEKYKAVS ISTEPPTYLR EQKAKRNSQW VPTLPNSSHH LDAVPCSTTI NRNRMGRDKK RTFPLCFDDH DPAVIHENAS QPEVLVPIRL DMEIDGQKLR DAFTWNMNEK LMTPEMFSEI LCDDLDLNPL TFVPAIASAI RQQIESYPTD SILEDQSDQR VIIKLNIHVG NISLVDQFEW DMSEKENSPE KFALKLCSEL GLGGEFVTTI AYSIRGQLSW HQKTYAFSEN PLPTVEIAIR NTGDADQWCP LLETLTDAEM EKKIRDQDRN TRRMR
Buffer Tris-based buffer, 50% glycerol
Purity or Quality Grade Greater than 90% as determined by SDS-PAGE.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.