Learn More
enQuireBio™ Recombinant Human Sclerostin domain-containing protein 1
A DNA sequence encoding the Homo sapiens (Human) Sclerostin domain-containing protein 1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6717-ec-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_056279.1 | |
Sclerostin domain-containing protein 1 | |
Research Use Only | |
SOSTDC1 | |
Recombinant Protein | |
E. coli | |
FKNDATEILY SHVVKPVPAH PSSNSTLNQA RNGGRHFSNT GLDRNTRVQV GCRELRSTKY ISDGQCTSIS PLKELVCAGE CLPLPVLPNW IGGGYGTKYW SRRSSQEWRC VNDKTRTQRI QLQCQDGSTR TYKITVVTAC KCKRYTRQHN ESSHNFESMS PAKPVQHHRE RKRASKSSKH SMS | |
Greater than 90% as determined by SDS-PAGE. |
25928 | |
500 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |