missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Sclerostin domain-containing protein 1
A DNA sequence encoding the Homo sapiens (Human) Sclerostin domain-containing protein 1, was expressed in the hosts and tags indicated.
160.00€ - 2385.00€
Specifications
Accession Number | NP_056279.1 |
---|---|
Gene ID (Entrez) | 25928 |
Name | Sclerostin domain-containing protein 1 |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16015350
|
enQuireBio™
QP6717-EC-100UG |
100 μg |
530.00€
100µg |
Estimated Shipment: 17-06-2024
Log in to see stock. |
|||||
16035350
|
enQuireBio™
QP6717-EC-1MG |
1 mg |
2385.00€
1mg |
Estimated Shipment: 17-06-2024
Log in to see stock. |
|||||
16025350
|
enQuireBio™
QP6717-EC-10UG |
10 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16045350
|
enQuireBio™
QP6717-EC-200UG |
200 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16055350
|
enQuireBio™
QP6717-EC-500UG |
500 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16065350
|
enQuireBio™
QP6717-EC-50UG |
50 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
Specifications
NP_056279.1 | |
Sclerostin domain-containing protein 1 | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |
25928 | |
Research Use Only | |
SOSTDC1 | |
Recombinant Protein | |
E. coli | |
FKNDATEILY SHVVKPVPAH PSSNSTLNQA RNGGRHFSNT GLDRNTRVQV GCRELRSTKY ISDGQCTSIS PLKELVCAGE CLPLPVLPNW IGGGYGTKYW SRRSSQEWRC VNDKTRTQRI QLQCQDGSTR TYKITVVTAC KCKRYTRQHN ESSHNFESMS PAKPVQHHRE RKRASKSSKH SMS | |
Greater than 90% as determined by SDS-PAGE. |