missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human SAP30-binding Protein
A DNA sequence encoding the Homo sapiens (Human) SAP30-binding protein, was expressed in the hosts and tags indicated.
160.00€ - 2385.00€
Specifications
Accession Number | NP_001288768.1 |
---|---|
Gene ID (Entrez) | 29115 |
Name | SAP30-binding Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16033141
|
enQuireBio™
QP8186-EC-100UG |
100 μg |
530.00€
100µg |
Estimated Shipment: 25-06-2024
Log in to see stock. |
|||||
16053141
|
enQuireBio™
QP8186-EC-1MG |
1 mg |
2385.00€
1mg |
Estimated Shipment: 25-06-2024
Log in to see stock. |
|||||
16043141
|
enQuireBio™
QP8186-EC-10UG |
10 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16063141
|
enQuireBio™
QP8186-EC-200UG |
200 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16073141
|
enQuireBio™
QP8186-EC-500UG |
500 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16083141
|
enQuireBio™
QP8186-EC-50UG |
50 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
Specifications
NP_001288768.1 | |
SAP30-binding Protein | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |
29115 | |
Research Use Only | |
SAP30BP | |
Recombinant Protein | |
E. coli | |
MAGKKNVLSS LAVYAEDSEP ESDGEAGIEA VGSAAEEKGG LVSDAYGEDD FSRLGGDEDG YEEEEDENSR QSEDDDSETE KPEADDPKDN TEAEKRDPQE LVASFSERVR NMSPDEIKIP PEPPGRCSNH LQDKIQKLYE RKIKEGMDMN YIIQRKKEFR NPSIYEKLIQ FCAIDELGTN YPKDMFDPHG WSEDSYYEAL AKAQKIEMDK LEKAKKERTK IEFVTGTKKG TTTNATSTTT TTASTAVADA QKRKSKWDSA IPVTTIAQPT ILTTTATLPA VVTVTTSASG SKTTVISAVG TIVKKAKQ | |
Greater than 80% as determined by SDS-PAGE. |