missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human SAP30-binding Protein

A DNA sequence encoding the Homo sapiens (Human) SAP30-binding protein, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8186-ec-50ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16083141

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance


Description

Description

Specifications

Specifications

NP_001288768.1
SAP30-binding Protein
Research Use Only
SAP30BP
Recombinant Protein
E. coli
MAGKKNVLSS LAVYAEDSEP ESDGEAGIEA VGSAAEEKGG LVSDAYGEDD FSRLGGDEDG YEEEEDENSR QSEDDDSETE KPEADDPKDN TEAEKRDPQE LVASFSERVR NMSPDEIKIP PEPPGRCSNH LQDKIQKLYE RKIKEGMDMN YIIQRKKEFR NPSIYEKLIQ FCAIDELGTN YPKDMFDPHG WSEDSYYEAL AKAQKIEMDK LEKAKKERTK IEFVTGTKKG TTTNATSTTT TTASTAVADA QKRKSKWDSA IPVTTIAQPT ILTTTATLPA VVTVTTSASG SKTTVISAVG TIVKKAKQ
Greater than 80% as determined by SDS-PAGE.
29115
50 μg
Not determined.
Full Length
Human
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers