missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human RBKS / Ribokinase Protein
A DNA sequence encoding the Homo sapiens (Human) RBKS / Ribokinase, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6585-ec-200ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001274509.1 | |
RBKS / Ribokinase Protein | |
Research Use Only | |
RBKS | |
Recombinant Protein | |
E. coli | |
AASGEPQRQW QEEVAAVVVV GSCMTDLVSL TSRLPKTGET IHGHKFFIGF GGKGANQCVQ AARLGAMTSM VCKVGKDSFG NDYIENLKQN DISTEFTYQT KDAATGTASI IVNNEGQNII VIVAGANLLL NTEDLRAAAN VISRAKVMVC QLEITPATSL EALTMARRSG VKTLFNPAPA IADLDPQFYT LSDVFCCNES EAEILTGLTV GSAADAGEAA LVLLKRGCQV VIITLGAEGC VVLSQTEPEP KHIPTEKVKA VDTTGAGDSF VGALAFYLAY YPNLSLEDML NRSNFIAAVS VQAAGTQSSY PYKKDLPLTL F | |
Greater than 90% as determined by SDS-PAGE. |
64080 | |
200 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |