missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human RBKS / Ribokinase Protein
A DNA sequence encoding the Homo sapiens (Human) RBKS / Ribokinase, was expressed in the hosts and tags indicated.
160.00€ - 2385.00€
Specifications
Accession Number | NP_001274509.1 |
---|---|
Gene ID (Entrez) | 64080 |
Name | RBKS / Ribokinase Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16004340
|
enQuireBio™
QP6585-EC-100UG |
100 μg |
530.00€
100µg |
Estimated Shipment: 21-06-2024
Log in to see stock. |
|||||
16024340
|
enQuireBio™
QP6585-EC-1MG |
1 mg |
2385.00€
1mg |
Estimated Shipment: 21-06-2024
Log in to see stock. |
|||||
16014340
|
enQuireBio™
QP6585-EC-10UG |
10 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16034340
|
enQuireBio™
QP6585-EC-200UG |
200 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16044340
|
enQuireBio™
QP6585-EC-500UG |
500 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16054340
|
enQuireBio™
QP6585-EC-50UG |
50 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
Specifications
NP_001274509.1 | |
RBKS / Ribokinase Protein | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
64080 | |
Research Use Only | |
RBKS | |
Recombinant Protein | |
E. coli | |
AASGEPQRQW QEEVAAVVVV GSCMTDLVSL TSRLPKTGET IHGHKFFIGF GGKGANQCVQ AARLGAMTSM VCKVGKDSFG NDYIENLKQN DISTEFTYQT KDAATGTASI IVNNEGQNII VIVAGANLLL NTEDLRAAAN VISRAKVMVC QLEITPATSL EALTMARRSG VKTLFNPAPA IADLDPQFYT LSDVFCCNES EAEILTGLTV GSAADAGEAA LVLLKRGCQV VIITLGAEGC VVLSQTEPEP KHIPTEKVKA VDTTGAGDSF VGALAFYLAY YPNLSLEDML NRSNFIAAVS VQAAGTQSSY PYKKDLPLTL F | |
Greater than 90% as determined by SDS-PAGE. |