missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Ras-related protein Rab-5C Protein
A DNA sequence encoding the Homo sapiens (Human) Ras-related protein Rab-5C, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6573-ec-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001238968.1 | |
Ras-related protein Rab-5C Protein | |
Research Use Only | |
RAB5C | |
Recombinant Protein | |
E. coli | |
MAGRGGAARP NGPAAGNKIC QFKLVLLGES AVGKSSLVLR FVKGQFHEYQ ESTIGAAFLT QTVCLDDTTV KFEIWDTAGQ ERYHSLAPMY YRGAQAAIVV YDITNTDTFA RAKNWVKELQ RQASPNIVIA LAGNKADLAS KRAVEFQEAQ AYADDNSLLF METSAKTAMN VNEIFMAIAK KLPKNEPQNA TGAPGRNRGV DLQENNPASR SQCCSN | |
Greater than 90% as determined by SDS-PAGE. |
5878 | |
50 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |