missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Ras-related protein Rab-5C Protein
A DNA sequence encoding the Homo sapiens (Human) Ras-related protein Rab-5C, was expressed in the hosts and tags indicated.
160.00€ - 2385.00€
Specifications
Accession Number | NP_001238968.1 |
---|---|
Gene ID (Entrez) | 5878 |
Name | Ras-related protein Rab-5C Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16024260
|
enQuireBio™
QP6573-EC-100UG |
100 μg |
530.00€
100µg |
Estimated Shipment: 17-06-2024
Log in to see stock. |
|||||
16044260
|
enQuireBio™
QP6573-EC-1MG |
1 mg |
2385.00€
1mg |
Estimated Shipment: 17-06-2024
Log in to see stock. |
|||||
16034260
|
enQuireBio™
QP6573-EC-10UG |
10 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16054260
|
enQuireBio™
QP6573-EC-200UG |
200 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16064260
|
enQuireBio™
QP6573-EC-500UG |
500 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16074260
|
enQuireBio™
QP6573-EC-50UG |
50 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
Specifications
NP_001238968.1 | |
Ras-related protein Rab-5C Protein | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
5878 | |
Research Use Only | |
RAB5C | |
Recombinant Protein | |
E. coli | |
MAGRGGAARP NGPAAGNKIC QFKLVLLGES AVGKSSLVLR FVKGQFHEYQ ESTIGAAFLT QTVCLDDTTV KFEIWDTAGQ ERYHSLAPMY YRGAQAAIVV YDITNTDTFA RAKNWVKELQ RQASPNIVIA LAGNKADLAS KRAVEFQEAQ AYADDNSLLF METSAKTAMN VNEIFMAIAK KLPKNEPQNA TGAPGRNRGV DLQENNPASR SQCCSN | |
Greater than 90% as determined by SDS-PAGE. |